Product Details

SNP ID
rs140574615
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:26994857 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAAGCATCTCCTACGACCTAGAC[A/G]GGGAAGAGAGCTATGGCAAGTACCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FIBIN PubMed Links

Gene Details

Gene
FIBIN
Gene Name
fin bud initiation factor homolog (zebrafish)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_203371.1 777 Missense Mutation AGG,GGG R111G NP_976249.1

View Full Product Details