Product Details

SNP ID
rs140687140
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:118660626 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGGCACCAGTTTGCTTGTTCGGA[C/T]GCCGCTAAGCGAGTTGGGGTTTGGG
Phenotype
MIM: 612834 MIM: 275360
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PHLDB1 PubMed Links

Gene Details

Gene
PHLDB1
Gene Name
pleckstrin homology like domain family B member 1
There are no transcripts associated with this gene.

Gene
TREH
Gene Name
trehalase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301065.1 1060 Missense Mutation ATC,GTC I308V NP_001287994.1
NM_007180.2 1060 Missense Mutation ATC,GTC I339V NP_009111.2

View Full Product Details