Product Details

SNP ID
rs140805031
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:73309702 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTCTCCGCCCTGCGTCCCAGGTC[C/T]CCAGGAGGGACTTCGGCCTATGTCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARHGEF17 PubMed Links

Gene Details

Gene
ARHGEF17
Gene Name
Rho guanine nucleotide exchange factor 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014786.3 1085 Missense Mutation CCC,CTC P355L NP_055601.2
XM_017018623.1 1085 Intron XP_016874112.1
XM_017018624.1 1085 Intron XP_016874113.1

View Full Product Details