Product Details

SNP ID
rs140964535
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:69699427 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGCAGCATGGGCAGGAAATGAGA[G/T]AGTGGAAGAAAGCCTCTGTTCTTGT
Phenotype
MIM: 603891
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FGF19 PubMed Links

Gene Details

Gene
FGF19
Gene Name
fibroblast growth factor 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005117.2 949 Silent Mutation CTA,CTC L162L NP_005108.1

View Full Product Details