Product Details

SNP ID
rs141278630
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:66000116 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCTCACCAGGAAGCGCTGCCCTT[C/G]GGCTGTCTCCACCTCATGCAGATTG
Phenotype
MIM: 603811
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
BANF1 PubMed Links

Gene Details

Gene
BANF1
Gene Name
barrier to autointegration factor 1
There are no transcripts associated with this gene.

Gene
EIF1AD
Gene Name
eukaryotic translation initiation factor 1A domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242481.1 277 Missense Mutation CAA,GAA Q45E NP_001229410.1
NM_001242482.1 277 Missense Mutation CAA,GAA Q45E NP_001229411.1
NM_001242483.1 277 Missense Mutation CAA,GAA Q45E NP_001229412.1
NM_001242484.1 277 Missense Mutation CAA,GAA Q45E NP_001229413.1
NM_001242485.1 277 Missense Mutation CAA,GAA Q45E NP_001229414.1
NM_001242486.1 277 Missense Mutation CAA,GAA Q45E NP_001229415.1
NM_032325.3 277 Missense Mutation CAA,GAA Q45E NP_115701.2
XM_017018412.1 277 Missense Mutation CAA,GAA Q45E XP_016873901.1

View Full Product Details