Product Details

SNP ID
rs141464502
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:65884450 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTGATTCAGGAAGAACCGCACGT[C/T]GCTGAGTGGCCAGTGGTCGGAGCGG
Phenotype
MIM: 605360 MIM: 602364 MIM: 608296 MIM: 136515
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC85B PubMed Links

Gene Details

Gene
CCDC85B
Gene Name
coiled-coil domain containing 85B
There are no transcripts associated with this gene.

Gene
CTSW
Gene Name
cathepsin W
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001335.3 1088 Intron NP_001326.2
Gene
FIBP
Gene Name
FGF1 intracellular binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004214.4 1088 Missense Mutation AAC,GAC N316D NP_004205.2
NM_198897.1 1088 Missense Mutation AAC,GAC N323D NP_942600.1
Gene
FOSL1
Gene Name
FOS like 1, AP-1 transcription factor subunit
There are no transcripts associated with this gene.

View Full Product Details