Product Details

SNP ID
rs141939785
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5787939 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCCTGGTTTTCACCCCATACACA[A/G]TAGGGTTCATTGTGGGAGGCATTAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR52N1 PubMed Links

Gene Details

Gene
OR52N1
Gene Name
olfactory receptor family 52 subfamily N member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001913.1 878 Missense Mutation ACT,ATT T293I NP_001001913.1
Gene
OR52N5
Gene Name
olfactory receptor family 52 subfamily N member 5
There are no transcripts associated with this gene.

View Full Product Details