Product Details

SNP ID
rs142474714
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:55819864 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATTTCTGCTATTCCTCCATCATT[G/T]CTCCCAAGATGTTGGTGAACCTTGT
Phenotype
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
OR5D18 PubMed Links
Additional Information
For this assay, SNP(s) [rs7948629] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
OR5D18
Gene Name
olfactory receptor family 5 subfamily D member 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001952.1 235 Missense Mutation GCT,TCT A79S NP_001001952.1
Gene
OR5L1
Gene Name
olfactory receptor family 5 subfamily L member 1 (gene/pseudogene)
There are no transcripts associated with this gene.

Gene
OR5L2
Gene Name
olfactory receptor family 5 subfamily L member 2
There are no transcripts associated with this gene.

View Full Product Details