Product Details

SNP ID
rs142623287
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:7938716 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCAATGAGATCAGTTTCTTGGTTT[C/T]GGGTGAGTAGCCAGATTTGGGTTGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR10A3 PubMed Links

Gene Details

Gene
OR10A3
Gene Name
olfactory receptor family 10 subfamily A member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003745.1 Intron NP_001003745.1

View Full Product Details