Product Details

SNP ID
rs142656646
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:73309641 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTGAGCCCCCAACATCTCCAAGA[A/G]CCCCTAGAGAAGAAGGACTCCGGGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARHGEF17 PubMed Links

Gene Details

Gene
ARHGEF17
Gene Name
Rho guanine nucleotide exchange factor 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014786.3 1024 Missense Mutation ACC,GCC T335A NP_055601.2
XM_017018623.1 1024 Intron XP_016874112.1
XM_017018624.1 1024 Intron XP_016874113.1

View Full Product Details