Product Details

SNP ID
rs142931568
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:130231403 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGCAGCAGACAACTAACCAGATA[C/T]GCATTTTCACAGTATATTCAGATTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZBTB44 PubMed Links

Gene Details

Gene
ZBTB44
Gene Name
zinc finger and BTB domain containing 44
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301098.1 4634 UTR 3 NP_001288027.1
NM_001301099.1 4634 UTR 3 NP_001288028.1
NM_014155.4 4634 UTR 3 NP_054874.3
XM_005271521.4 4634 UTR 3 XP_005271578.1
XM_005271522.4 4634 UTR 3 XP_005271579.1
XM_006718825.3 4634 UTR 3 XP_006718888.1
XM_006718826.3 4634 UTR 3 XP_006718889.1
XM_011542789.2 4634 Intron XP_011541091.1
XM_017017623.1 4634 UTR 3 XP_016873112.1
XM_017017624.1 4634 UTR 3 XP_016873113.1

View Full Product Details