Product Details

SNP ID
rs142947187
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:61789892 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGAGGAGCTCTTTATAGATATCA[C/T]GAGTGTACTTGGTAGAGGTGACCTC
Phenotype
MIM: 606148 MIM: 600393 MIM: 608329
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FADS1 PubMed Links

Gene Details

Gene
FADS1
Gene Name
fatty acid desaturase 1
There are no transcripts associated with this gene.

Gene
FEN1
Gene Name
flap structure-specific endonuclease 1
There are no transcripts associated with this gene.

Gene
MIR611
Gene Name
microRNA 611
There are no transcripts associated with this gene.

Gene
MYRF
Gene Name
myelin regulatory factor
There are no transcripts associated with this gene.

Gene
TMEM258
Gene Name
transmembrane protein 258
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014206.3 Intron NP_055021.1

View Full Product Details