Product Details

SNP ID
rs142969856
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5132066 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATATCTTGTTGACTCGGATATCTT[C/T]AGTAGCCAGCTTCACGATGGCCATG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR52A5 PubMed Links

Gene Details

Gene
OR52A5
Gene Name
olfactory receptor family 52 subfamily A member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005160.2 577 Missense Mutation AAA,GAA K193E NP_001005160.1

View Full Product Details