Product Details

SNP ID
rs143434418
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:14644958 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTGTGATCCGACCCCGGAGGAGG[C/T]CCAGCTGCGTGTCGTTAGGAGAAAC
Phenotype
MIM: 602047 MIM: 602854
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PDE3B PubMed Links

Gene Details

Gene
PDE3B
Gene Name
phosphodiesterase 3B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000922.3 1268 Missense Mutation CCC,TCC P295S NP_000913.2
XM_006718249.3 1268 Missense Mutation CCC,TCC P295S XP_006718312.1
XM_017017911.1 1268 Missense Mutation CCC,TCC P295S XP_016873400.1
XM_017017912.1 1268 Missense Mutation CCC,TCC P295S XP_016873401.1
Gene
PSMA1
Gene Name
proteasome subunit alpha 1
There are no transcripts associated with this gene.

View Full Product Details