Product Details

SNP ID
rs143982811
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:65920032 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGCGGAACGCGAAGACCATGACCA[C/T]ATCCCACCTGTGAGCGGCGAGGAGC
Phenotype
MIM: 602289
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C11orf68 PubMed Links

Gene Details

Gene
C11orf68
Gene Name
chromosome 11 open reading frame 68
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135635.1 445 Intron NP_001129107.1
NM_031450.3 445 Intron NP_113638.2
Gene
DRAP1
Gene Name
DR1 associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006442.3 445 Missense Mutation ACA,ATA T67I NP_006433.2

View Full Product Details