Product Details

SNP ID
rs144053540
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:6026892 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATGTCCATGGGGACCTTCTTTCT[A/G]GCCACGTTTGTCAACACCACAACCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR56A1 PubMed Links

Gene Details

Gene
OR56A1
Gene Name
olfactory receptor family 56 subfamily A member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001917.2 850 Silent Mutation GCC,GCT A271A NP_001001917.2

View Full Product Details