Product Details

SNP ID
rs144928396
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:418531 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCTCCAGAACCTTGTTCTTCACC[A/G]ACTGAGGGATGTCGGGCACGAACCA
Phenotype
MIM: 605478
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANO9 PubMed Links

Gene Details

Gene
ANO9
Gene Name
anoctamin 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012302.2 2274 Missense Mutation TCG,TTG S730L NP_001012302.2
XM_011520048.1 2274 Intron XP_011518350.1
XM_011520051.2 2274 Intron XP_011518353.1
XM_011520052.2 2274 Intron XP_011518354.1
XM_011520053.2 2274 Missense Mutation TCG,TTG S431L XP_011518355.1
XM_017017644.1 2274 Intron XP_016873133.1
XM_017017645.1 2274 Intron XP_016873134.1
XM_017017646.1 2274 Intron XP_016873135.1
XM_017017647.1 2274 Intron XP_016873136.1
Gene
SIGIRR
Gene Name
single Ig and TIR domain containing
There are no transcripts associated with this gene.

View Full Product Details