Product Details

SNP ID
rs144963209
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5323643 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAATGCCTATGACAGTATTAAGAA[G/T]TAGAATATAGGAGAAGAGGATGATC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OR51B2 PubMed Links

Gene Details

Gene
OR51B2
Gene Name
olfactory receptor family 51 subfamily B member 2 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033180.4 710 Missense Mutation ATT,CTT I219L NP_149420.4

View Full Product Details