Product Details
- SNP ID
-
rs145107371
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:72110109 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTACCTCATTGACCTCTCCATCATC[C/T]GGTGACTCATTGTAGTCATTCATCT
- Phenotype
-
MIM: 614717
MIM: 613510
MIM: 612414
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ANAPC15
PubMed Links
Gene Details
- Gene
- ANAPC15
- Gene Name
- anaphase promoting complex subunit 15
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001278485.1 |
430 |
Intron |
|
|
NP_001265414.1 |
NM_001278486.1 |
430 |
Intron |
|
|
NP_001265415.1 |
NM_001278487.1 |
430 |
Intron |
|
|
NP_001265416.1 |
NM_001278488.1 |
430 |
Silent Mutation |
CCA,CCG |
P99P |
NP_001265417.1 |
NM_001278489.1 |
430 |
Intron |
|
|
NP_001265418.1 |
NM_001278490.1 |
430 |
Intron |
|
|
NP_001265419.1 |
NM_001278491.1 |
430 |
Silent Mutation |
CCA,CCG |
P99P |
NP_001265420.1 |
NM_001278492.1 |
430 |
Silent Mutation |
CCA,CCG |
P99P |
NP_001265421.1 |
NM_001278493.1 |
430 |
Silent Mutation |
CCA,CCG |
P99P |
NP_001265422.1 |
NM_001278494.1 |
430 |
Silent Mutation |
CCA,CCG |
P99P |
NP_001265423.1 |
NM_014042.2 |
430 |
Intron |
|
|
NP_054761.1 |
XM_017017488.1 |
430 |
Silent Mutation |
CCA,CCG |
P99P |
XP_016872977.1 |
XM_017017489.1 |
430 |
Silent Mutation |
CCA,CCG |
P99P |
XP_016872978.1 |
XM_017017490.1 |
430 |
Silent Mutation |
CCA,CCG |
P99P |
XP_016872979.1 |
XM_017017491.1 |
430 |
Intron |
|
|
XP_016872980.1 |
XM_017017492.1 |
430 |
Silent Mutation |
CCA,CCG |
P99P |
XP_016872981.1 |
XM_017017493.1 |
430 |
Intron |
|
|
XP_016872982.1 |
XM_017017494.1 |
430 |
Silent Mutation |
CCA,CCG |
P99P |
XP_016872983.1 |
XM_017017495.1 |
430 |
Silent Mutation |
CCA,CCG |
P90P |
XP_016872984.1 |
XM_017017496.1 |
430 |
Silent Mutation |
CCA,CCG |
P99P |
XP_016872985.1 |
XM_017017497.1 |
430 |
Silent Mutation |
CCA,CCG |
P99P |
XP_016872986.1 |
- Gene
- LAMTOR1
- Gene Name
- late endosomal/lysosomal adaptor, MAPK and MTOR activator 1
There are no transcripts associated with this gene.
- Gene
- LRTOMT
- Gene Name
- leucine rich transmembrane and O-methyltransferase domain containing
View Full Product Details