Product Details

SNP ID
rs145283501
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:56641876 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAGCAAGAGAGTTTGAGCAGTGG[C/T]GGGGTGTCACAGTAGAAATGGTTGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR5AP2 PubMed Links

Gene Details

Gene
OR5AP2
Gene Name
olfactory receptor family 5 subfamily AP member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002925.1 564 Silent Mutation CCA,CCG P188P NP_001002925.1

View Full Product Details