Product Details

SNP ID
rs145377551
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:59515063 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCCTCATCATGGTTACAGTTACC[C/T]GTGAATCTCACCTTCATACGCCCAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR4D9 PubMed Links

Gene Details

Gene
OR4D9
Gene Name
olfactory receptor family 4 subfamily D member 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004711.1 151 Missense Mutation CGT,TGT R51C NP_001004711.1

View Full Product Details