Product Details

SNP ID
rs145616071
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:55914057 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGGAGGGATATCACAGAAGAAAT[A/G]ATTAATCTCATTAGACCCACAGAAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR5W2 PubMed Links

Gene Details

Gene
OR5W2
Gene Name
olfactory receptor family 5 subfamily W member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001960.1 526 Missense Mutation CAT,TAT H176Y NP_001001960.1

View Full Product Details