Product Details
- SNP ID
-
rs145655133
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:5301530 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCATCAGTACCCCCAGTCCTATGTT[A/C]ATCACTCGGGAATTGGTAAGAATGC
- Phenotype
-
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
OR51B4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs10837771] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OR51B4
- Gene Name
- olfactory receptor family 51 subfamily B member 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_033179.2 |
417 |
Missense Mutation |
|
|
NP_149419.2 |
View Full Product Details