Product Details

SNP ID
rs145687206
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:22341610 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCCTAGAGGTGCCCGAGAGGAAG[G/T]CGCCGCTGTGCGACTGCACGTGCTT
Phenotype
MIM: 607563
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SLC17A6 PubMed Links

Gene Details

Gene
SLC17A6
Gene Name
solute carrier family 17 member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020346.2 582 Missense Mutation GCG,TCG A57S NP_065079.1

View Full Product Details