Product Details
- SNP ID
-
rs146398167
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:60173059 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAAGTCAGAGTAAGCCTGTTTCCAC[C/T]GCAGCACAGCAGTGAGCACAGCTAG
- Phenotype
-
MIM: 606548
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MS4A6A
PubMed Links
Gene Details
- Gene
- MS4A6A
- Gene Name
- membrane spanning 4-domains A6A
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001247999.1 |
884 |
Missense Mutation |
CAG,CGG |
Q235R |
NP_001234928.1 |
NM_022349.3 |
884 |
Missense Mutation |
CAG,CGG |
Q207R |
NP_071744.2 |
NM_152851.2 |
884 |
Silent Mutation |
GCA,GCG |
A172A |
NP_690590.1 |
NM_152852.2 |
884 |
Missense Mutation |
CAG,CGG |
Q207R |
NP_690591.1 |
XM_005274177.2 |
884 |
Missense Mutation |
CAG,CGG |
Q235R |
XP_005274234.1 |
XM_006718658.2 |
884 |
Missense Mutation |
CAG,CGG |
Q235R |
XP_006718721.1 |
XM_006718660.2 |
884 |
Missense Mutation |
CAG,CGG |
Q190R |
XP_006718723.1 |
XM_006718661.2 |
884 |
Missense Mutation |
CAG,CGG |
Q190R |
XP_006718724.1 |
XM_011545209.1 |
884 |
Intron |
|
|
XP_011543511.1 |
XM_017018123.1 |
884 |
Missense Mutation |
CAG,CGG |
Q162R |
XP_016873612.1 |
XM_017018124.1 |
884 |
Missense Mutation |
CAG,CGG |
Q162R |
XP_016873613.1 |
XM_017018125.1 |
884 |
Intron |
|
|
XP_016873614.1 |
View Full Product Details