Product Details

SNP ID
rs146411780
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:66332580 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGGCCAGGGCCCCAGGGGGCAGG[C/T]GGTGGTAGCCCTGCTCCTTGTACAG
Phenotype
MIM: 606259 MIM: 605965
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BRMS1 PubMed Links

Gene Details

Gene
BRMS1
Gene Name
breast cancer metastasis suppressor 1
There are no transcripts associated with this gene.

Gene
RIN1
Gene Name
Ras and Rab interactor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004292.2 2524 Missense Mutation CAC,CGC H683R NP_004283.2
XM_011545399.2 2524 Missense Mutation CAC,CGC H655R XP_011543701.1
XM_011545400.2 2524 Missense Mutation CAC,CGC H621R XP_011543702.1
XM_011545401.2 2524 Missense Mutation CAC,CGC H602R XP_011543703.1
XM_017018587.1 2524 Missense Mutation CAC,CGC H622R XP_016874076.1

View Full Product Details