Product Details

SNP ID
rs146560356
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:60891214 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTGTCCATGCCTGTTGAAGCGAT[G/T]AACTTGGCGTGATGCCCGAAGGCCA
Phenotype
MIM: 608330
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PRPF19 PubMed Links

Gene Details

Gene
PRPF19
Gene Name
pre-mRNA processing factor 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014502.4 1674 Missense Mutation TTA,TTC L489F NP_055317.1

View Full Product Details