Product Details

SNP ID
rs146869210
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:55914041 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGAGAGAGTAATAAGAGAGGAGGG[A/G]TATCACAGAAGAAATGATTAATCTC
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
OR5W2 PubMed Links

Gene Details

Gene
OR5W2
Gene Name
olfactory receptor family 5 subfamily W member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001960.1 542 Missense Mutation ACC,ATC T181I NP_001001960.1

View Full Product Details