Product Details

SNP ID
rs146971652
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:119339670 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACGGCGTCGTAATGTCCCTGCTC[G/T]TTCACCAGCACGCGGTCGAAGGGCA
Phenotype
MIM: 608752 MIM: 606227 MIM: 606130
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C1QTNF5 PubMed Links

Gene Details

Gene
C1QTNF5
Gene Name
C1q and tumor necrosis factor related protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278431.1 625 Missense Mutation AAA,AAC K131N NP_001265360.1
NM_015645.4 625 Missense Mutation AAA,AAC K131N NP_056460.1
Gene
MFRP
Gene Name
membrane frizzled-related protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031433.3 625 UTR 3 NP_113621.1
Gene
RNF26
Gene Name
ring finger protein 26
There are no transcripts associated with this gene.

View Full Product Details