Product Details

SNP ID
rs146972287
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:6199603 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGCTTTTGGCCATGGCCTGTGATC[A/G]TGCTGCGGCAATAGGGCGTCCACTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C11orf42 PubMed Links

Gene Details

Gene
C11orf42
Gene Name
chromosome 11 open reading frame 42
There are no transcripts associated with this gene.

Gene
OR52W1
Gene Name
olfactory receptor family 52 subfamily W member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005178.1 380 Missense Mutation CAT,CGT H127R NP_001005178.1

View Full Product Details