Product Details

SNP ID
rs147044490
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:123976872 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGCCTCACTGGAGCGAGGCTGCA[A/G]GTAGATACAGACAGGTGGCACGTAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR10S1 PubMed Links

Gene Details

Gene
OR10S1
Gene Name
olfactory receptor family 10 subfamily S member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004474.1 820 Silent Mutation CTG,TTG L274L NP_001004474.1

View Full Product Details