Product Details

SNP ID
rs147226965
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:85698040 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGGTAGATCAAGGCATTCCTTCA[A/C]CCAGATGTGCACTTCTCCAGTTGTA
Phenotype
MIM: 612880
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SYTL2 PubMed Links

Gene Details

Gene
SYTL2
Gene Name
synaptotagmin like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001162951.2 6375 Missense Mutation NP_001156423.1
NM_001162952.2 6375 Missense Mutation NP_001156424.1
NM_001162953.2 6375 Missense Mutation NP_001156425.1
NM_001289608.1 6375 Missense Mutation NP_001276537.1
NM_001289609.1 6375 Missense Mutation NP_001276538.1
NM_001289610.1 6375 Missense Mutation NP_001276539.1
NM_032943.4 6375 Missense Mutation NP_116561.1
NM_206929.3 6375 Missense Mutation NP_996812.1
NM_206930.3 6375 Missense Mutation NP_996813.1
XM_005274057.1 6375 Missense Mutation XP_005274114.1
XM_005274060.1 6375 Missense Mutation XP_005274117.1
XM_005274061.1 6375 Missense Mutation XP_005274118.1
XM_005274062.2 6375 Missense Mutation XP_005274119.1
XM_005274063.1 6375 Missense Mutation XP_005274120.1
XM_005274066.1 6375 Missense Mutation XP_005274123.1
XM_005274067.4 6375 Missense Mutation XP_005274124.1
XM_011545105.1 6375 Missense Mutation XP_011543407.1
XM_011545106.1 6375 Missense Mutation XP_011543408.1
XM_011545107.2 6375 Missense Mutation XP_011543409.1
XM_011545108.1 6375 Missense Mutation XP_011543410.1
XM_011545109.2 6375 Missense Mutation XP_011543411.1
XM_011545110.1 6375 Missense Mutation XP_011543412.1
XM_011545112.1 6375 Intron XP_011543414.1
XM_011545114.2 6375 Missense Mutation XP_011543416.1
XM_011545115.1 6375 Missense Mutation XP_011543417.1
XM_011545116.2 6375 Missense Mutation XP_011543418.1
XM_017017933.1 6375 Missense Mutation XP_016873422.1
XM_017017934.1 6375 Missense Mutation XP_016873423.1
XM_017017935.1 6375 Missense Mutation XP_016873424.1
XM_017017936.1 6375 Missense Mutation XP_016873425.1
XM_017017937.1 6375 Missense Mutation XP_016873426.1
XM_017017938.1 6375 Missense Mutation XP_016873427.1
XM_017017939.1 6375 Missense Mutation XP_016873428.1
XM_017017940.1 6375 Missense Mutation XP_016873429.1

View Full Product Details