Product Details

SNP ID
rs147268880
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:6498809 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCTGAGCTACAGCGCTGCCTGGG[A/C]ATTGTTGGTGCTCAGGTGGCCCTAG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DNHD1 PubMed Links

Gene Details

Gene
DNHD1
Gene Name
dynein heavy chain domain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144666.2 1158 Silent Mutation GGA,GGC G198G NP_653267.2
NM_173589.3 1158 Silent Mutation GGA,GGC G198G NP_775860.3

View Full Product Details