Product Details

SNP ID
rs147895623
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:49981933 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAACAAAGAGACCAAGGGTATGA[A/G]TGTCTATGCAAACAAGTTTTAACAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR4C12 PubMed Links

Gene Details

Gene
OR4C12
Gene Name
olfactory receptor family 4 subfamily C member 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005270.4 Intron NP_001005270.3

View Full Product Details