Product Details

SNP ID
rs148138297
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:68261995 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGATGTGCCTGGCCCAGCGGCCC[A/T]CTGGGTATATGGCATGGGGCTTGGC
Phenotype
MIM: 610880
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
C11orf24 PubMed Links

Gene Details

Gene
C11orf24
Gene Name
chromosome 11 open reading frame 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300913.1 1442 Intron NP_001287842.1
NM_022338.3 1442 Missense Mutation AGG,TGG R334W NP_071733.1
XM_005274053.3 1442 Missense Mutation AGG,TGG R334W XP_005274110.1
XM_011545103.2 1442 Missense Mutation AGG,TGG R302W XP_011543405.1

View Full Product Details