Product Details

SNP ID
rs148233159
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:64895018 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCGCCACTTGAGGGCGTGGTCCTT[A/G]TGGGCGTCGGGGACAATCTGGTTGC
Phenotype
MIM: 616225 MIM: 610939 MIM: 610941
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATG2A PubMed Links

Gene Details

Gene
ATG2A
Gene Name
autophagy related 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015104.2 3498 Silent Mutation CAC,CAT H1924H NP_055919.2
XM_005273849.3 3498 Silent Mutation CAC,CAT H1918H XP_005273906.1
XM_005273850.3 3498 Silent Mutation CAC,CAT H1916H XP_005273907.1
XM_011544863.2 3498 Silent Mutation CAC,CAT H1926H XP_011543165.1
XM_011544864.2 3498 Silent Mutation CAC,CAT H1921H XP_011543166.1
XM_011544865.2 3498 Silent Mutation CAC,CAT H1861H XP_011543167.1
XM_011544866.2 3498 Intron XP_011543168.1
XM_011544867.2 3498 Intron XP_011543169.1
XM_011544868.2 3498 Silent Mutation CAC,CAT H1138H XP_011543170.1
Gene
MIR192
Gene Name
microRNA 192
There are no transcripts associated with this gene.

Gene
MIR194-2
Gene Name
microRNA 194-2
There are no transcripts associated with this gene.

Gene
MIR194-2HG
Gene Name
MIR194-2 host gene
There are no transcripts associated with this gene.

Gene
MIR6749
Gene Name
microRNA 6749
There are no transcripts associated with this gene.

Gene
MIR6750
Gene Name
microRNA 6750
There are no transcripts associated with this gene.

View Full Product Details