Product Details

SNP ID
rs148495702
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:59854629 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAGAAATGACAGCAAACATCTTAA[C/G]CTTAGGTTAGGTACAGACCTACCCA
Phenotype
MIM: 609342 MIM: 189905
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GIF PubMed Links

Gene Details

Gene
GIF
Gene Name
gastric intrinsic factor
There are no transcripts associated with this gene.

Gene
TCN1
Gene Name
transcobalamin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001062.3 Intron NP_001053.2

View Full Product Details