Product Details

SNP ID
rs148709037
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:120516706 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGACATTTTCAAGAACAGCAGAGC[A/G]CGCTGACAGATTGGATGCAGCAGAT
Phenotype
MIM: 600282
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GRIK4 PubMed Links

Gene Details

Gene
GRIK4
Gene Name
glutamate ionotropic receptor kainate type subunit 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282470.2 Intron NP_001269399.1
NM_001282473.2 Intron NP_001269402.1
NM_014619.4 Intron NP_055434.2
XM_011542784.1 Intron XP_011541086.1
XM_011542786.2 Intron XP_011541088.1
XM_011542787.2 Intron XP_011541089.1
XM_017017621.1 Intron XP_016873110.1
XM_017017622.1 Intron XP_016873111.1

View Full Product Details