Product Details

SNP ID
rs148752326
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:26995798 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGATTTCTGATGCCTCTGTGGGT[A/G]TGTGTAAGTTAAAGTGGCCACATTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FIBIN PubMed Links

Gene Details

Gene
FIBIN
Gene Name
fin bud initiation factor homolog (zebrafish)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_203371.1 1718 UTR 3 NP_976249.1

View Full Product Details