Product Details

SNP ID
rs148997299
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:18701607 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTTGCTGTGACCCACATGTTCTA[C/T]GCCTCGGCCTTTGGCATGCAGCCAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IGSF22 PubMed Links

Gene Details

Gene
IGSF22
Gene Name
immunoglobulin superfamily member 22
There are no transcripts associated with this gene.

Gene
TMEM86A
Gene Name
transmembrane protein 86A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153347.2 473 Silent Mutation TAC,TAT Y107Y NP_699178.1

View Full Product Details