Product Details

SNP ID
rs149234091
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:129865951 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTCATTGGCTGGCTGATCACAGA[A/G]AGGGGAACTGTGATCCGTGTAGGCA
Phenotype
MIM: 164013
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NFRKB PubMed Links

Gene Details

Gene
NFRKB
Gene Name
nuclear factor related to kappaB binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143835.1 3676 Silent Mutation NP_001137307.1
NM_006165.3 3676 Silent Mutation NP_006156.2
XM_005271567.3 3676 Silent Mutation XP_005271624.1
XM_005271568.3 3676 Silent Mutation XP_005271625.1
XM_011542850.2 3676 Silent Mutation XP_011541152.1
XM_011542851.2 3676 Silent Mutation XP_011541153.1
XM_011542852.2 3676 Silent Mutation XP_011541154.1
XM_017017793.1 3676 Silent Mutation XP_016873282.1
XM_017017794.1 3676 Silent Mutation XP_016873283.1
XM_017017795.1 3676 Silent Mutation XP_016873284.1
XM_017017796.1 3676 Silent Mutation XP_016873285.1
XM_017017797.1 3676 Silent Mutation XP_016873286.1
XM_017017798.1 3676 Silent Mutation XP_016873287.1
XM_017017799.1 3676 Silent Mutation XP_016873288.1
XM_017017800.1 3676 Silent Mutation XP_016873289.1
XM_017017801.1 3676 Silent Mutation XP_016873290.1
Gene
TMEM45B
Gene Name
transmembrane protein 45B
There are no transcripts associated with this gene.

View Full Product Details