Product Details

SNP ID
rs149251939
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:48245434 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGATTTTCCTGCTCACTGTGATG[G/T]CCTATGACCACTATGTGGCCATCTG
Phenotype
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
OR4X2 PubMed Links

Gene Details

Gene
OR4X2
Gene Name
olfactory receptor family 4 subfamily X member 2 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004727.1 331 Missense Mutation GCC,TCC A111S NP_001004727.1

View Full Product Details