Product Details

SNP ID
rs149260337
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:61828489 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATTCAGAAGCATAACCTGCGCAC[C/T]GACAGGTGGCTGGTCATTGACCGCA
Phenotype
MIM: 606149
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FADS2 PubMed Links

Gene Details

Gene
FADS2
Gene Name
fatty acid desaturase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281501.1 457 Intron NP_001268430.1
NM_001281502.1 457 Intron NP_001268431.1
NM_004265.3 457 Silent Mutation ACC,ACT T33T NP_004256.1

View Full Product Details