Product Details

SNP ID
rs149354469
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:124424238 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCAAAGCGTAAGAGATGACGATGC[G/T]TATGCTGGATAGCATGGTGATTACT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OR8B4 PubMed Links

Gene Details

Gene
OR8B4
Gene Name
olfactory receptor family 8 subfamily B member 4 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005196.1 634 Missense Mutation AGC,CGC S212R NP_001005196.1

View Full Product Details