Product Details

SNP ID
rs149376071
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:67292109 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCCGCCATGCGGCCCAGCGAGGA[A/G]CATGTGGCCAGTCGCCTCACCTCTC
Phenotype
MIM: 615126 MIM: 109635
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANKRD13D PubMed Links

Gene Details

Gene
ANKRD13D
Gene Name
ankyrin repeat domain 13D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207354.2 93 Silent Mutation GAA,GAG E220E NP_997237.2
XM_011544985.1 93 Silent Mutation GAA,GAG E220E XP_011543287.1
XM_011544986.1 93 Silent Mutation GAA,GAG E133E XP_011543288.1
XM_011544987.2 93 Silent Mutation GAA,GAG E133E XP_011543289.1
XM_011544988.2 93 UTR 5 XP_011543290.1
Gene
GRK2
Gene Name
G protein-coupled receptor kinase 2
There are no transcripts associated with this gene.

View Full Product Details