Product Details

SNP ID
rs149591762
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:3666266 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAGGTATTGGCAATGGTGGCTAC[A/G]TGGTGCAGTAGGGCTTCCTGGCGGC
Phenotype
MIM: 601625 MIM: 606372 MIM: 601021
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ART1 PubMed Links

Gene Details

Gene
ART1
Gene Name
ADP-ribosyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004314.2 4462 Intron NP_004305.2
XM_011520114.2 4462 Intron XP_011518416.1
XM_017017763.1 4462 Intron XP_016873252.1
Gene
CHRNA10
Gene Name
cholinergic receptor nicotinic alpha 10 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303034.1 4462 Silent Mutation CAC,CAT H192H NP_001289963.1
NM_001303035.1 4462 Silent Mutation CAC,CAT H192H NP_001289964.1
NM_020402.3 4462 Silent Mutation CAC,CAT H398H NP_065135.2
XM_011520234.1 4462 Silent Mutation CAC,CAT H192H XP_011518536.1
Gene
NUP98
Gene Name
nucleoporin 98
There are no transcripts associated with this gene.

View Full Product Details