Product Details
- SNP ID
-
rs149649414
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:69810627 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGCGTACTAGACACCGTCCGGTAC[A/T]GCCGGGAGGCATACGTATTATAGCC
- Phenotype
-
MIM: 164950
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
FGF3
PubMed Links
Gene Details
- Gene
- FGF3
- Gene Name
- fibroblast growth factor 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005247.2 |
889 |
Missense Mutation |
CAG,CTG |
Q133L |
NP_005238.1 |
View Full Product Details