Product Details
- SNP ID
-
rs149702220
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:125097639 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGTGCAGTGGTTTGGCATAGATCGG[C/T]TCCAGAACATAATGGATTAAAACCA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SLC37A2
PubMed Links
Gene Details
- Gene
- SLC37A2
- Gene Name
- solute carrier family 37 member 2
There are no transcripts associated with this gene.
- Gene
- TMEM218
- Gene Name
- transmembrane protein 218
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001080546.2 |
947 |
Silent Mutation |
GAA,GAG |
E105E |
NP_001074015.1 |
NM_001258238.1 |
947 |
Silent Mutation |
GAA,GAG |
E105E |
NP_001245167.1 |
NM_001258239.1 |
947 |
Silent Mutation |
GAA,GAG |
E140E |
NP_001245168.1 |
NM_001258240.1 |
947 |
Silent Mutation |
GAA,GAG |
E105E |
NP_001245169.1 |
NM_001258241.1 |
947 |
Silent Mutation |
GAA,GAG |
E105E |
NP_001245170.1 |
NM_001258242.1 |
947 |
Silent Mutation |
GAA,GAG |
E140E |
NP_001245171.1 |
NM_001258243.1 |
947 |
Silent Mutation |
GAA,GAG |
E140E |
NP_001245172.1 |
NM_001258244.1 |
947 |
Silent Mutation |
GAA,GAG |
E105E |
NP_001245173.1 |
NM_001258245.1 |
947 |
Silent Mutation |
GAA,GAG |
E105E |
NP_001245174.1 |
NM_001258246.1 |
947 |
Silent Mutation |
GAA,GAG |
E140E |
NP_001245175.1 |
NM_001258247.1 |
947 |
Silent Mutation |
GAA,GAG |
E105E |
NP_001245176.1 |
XM_006718779.3 |
947 |
Silent Mutation |
GAA,GAG |
E140E |
XP_006718842.1 |
XM_006718780.3 |
947 |
Silent Mutation |
GAA,GAG |
E140E |
XP_006718843.1 |
XM_006718781.3 |
947 |
Silent Mutation |
GAA,GAG |
E140E |
XP_006718844.1 |
XM_006718783.3 |
947 |
Silent Mutation |
GAA,GAG |
E105E |
XP_006718846.1 |
XM_017017322.1 |
947 |
Silent Mutation |
GAA,GAG |
E105E |
XP_016872811.1 |
XM_017017323.1 |
947 |
Silent Mutation |
GAA,GAG |
E105E |
XP_016872812.1 |
XM_017017324.1 |
947 |
Silent Mutation |
GAA,GAG |
E105E |
XP_016872813.1 |
View Full Product Details