Product Details
- SNP ID
-
rs149782549
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:124922760 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGAGCTCACCTTCTGGTTTCAGGC[A/G]GTCATCATTCTGATCCATGTATTCC
- Phenotype
-
MIM: 611642
MIM: 611641
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
HEPACAM
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs11826299,rs77759828] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HEPACAM
- Gene Name
- hepatic and glial cell adhesion molecule
- Gene
- HEPN1
- Gene Name
- hepatocellular carcinoma, down-regulated 1
There are no transcripts associated with this gene.
View Full Product Details