Product Details

SNP ID
rs149927551
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:62909729 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAGAGGGGACTATCAGCATTGGC[A/G]GGAGGGAGTGCGGTGCACGGAGCCA
Phenotype
MIM: 118510
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CHRM1 PubMed Links

Gene Details

Gene
CHRM1
Gene Name
cholinergic receptor muscarinic 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000738.2 1474 Missense Mutation CGC,TGC R458C NP_000729.2
XM_011544742.2 1474 Missense Mutation CGC,TGC R458C XP_011543044.1

View Full Product Details